Calcium hydrogen orthophosphate dihydrate ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Calcium phosphate dibasic dihydrate, Calcium hydrogen phosphate dihydrate, Calcium hydrogenphosphate dihydrate, Dibasic Calcium Phosphate Dihydrate
MDL: MFCD00149621
Slightly soluble in water. Soluble in dilute hydrochloric, nitric, and acetic acid. Insoluble in alcohol
Purity: 98% min
CAS No.: 7789-77-7
Molecular Formula: CaHO4P.2H2O
Formula Weight: 172.09 (136.06anhy)
Physical Form: Powder
Density: 2.306
Appearance: White
Odor: Odorless
Melting Point: 109° -H2O
Solubility: Slightly soluble in water. Soluble in dilute hydrochloric, nitric, and acetic acid. Insoluble in alcohol
Merck Reference: 14,1692
MDL No.: MFCD00149621
Formula:
HCaO₄P·2H₂O MW: 172.09 g/mol Melting Pt: 109 °C (-H₂O) Density: 2.306 Storage Temperature: Ambient |
MDL Number:
MFCD00149621 CAS Number: 7789-77-7 EINECS: 231-826-1 Merck Index: 14,1692 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...