Calcium hydrogen orthophosphate, anhydrous ≥98%

Supplier: Thermo Scientific Chemicals

40232-30 40232-A1 40232-A7
CAAA40232-30EA 64.15 CAD
CAAA40232-30 CAAA40232-A1 CAAA40232-A7
Calcium hydrogen orthophosphate, anhydrous ≥98%
Calcium hydrogen orthophosphate

MDL: MFCD00010909


Purity: 98% min CAS No.: 7757-93-9 Molecular Formula: CaHO4P Formula Weight: 136.06 Physical Form: Powder Appearance: White Odor: Odorless Merck Reference: 14,1692 MDL No.: MFCD00010909

Formula: CaHPO₄
Melting Pt: 370 °C
Density: 2.89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010909
CAS Number: 7757-93-9
EINECS: 231-826-1
Merck Index: 14,1692

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR