Scandium ≥99.9% (metals basis excluding Ta), ingot (max. 8-12% Ta)
Supplier: Thermo Scientific Chemicals
40229-03
40229-06
CAAA40229-03EA
350.76
CAD
CAAA40229-03
CAAA40229-06
Scandium ≥99.9% (metals basis excluding Ta), ingot (max. 8-12% Ta)
Scandium
MDL: MFCD00016323
Purity: 99.9%
CAS No.: 7440-20-2
Physical Form: Packaged under argon
Appearance: Grey or silvery
Odor: Odorless
MDL No.: MFCD00016323
Formula:
Sc Boiling Pt: 2730 °C (1013 hPa) Melting Pt: 1541 °C Density: 2.989 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00016323 CAS Number: 7440-20-2 EINECS: 231-129-2 Merck Index: 12,08537 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...