Cobalt(II) bromide hydrate
Supplier: Thermo Scientific Chemicals
|
Warning
|
40225-18
40225-30
CAAA40225-18EA
130.19
CAD
CAAA40225-18
CAAA40225-30
Cobalt(II) bromide hydrate
Cobalt (II) bromide hydrate
Hygrometers, catalysts
CAS No.: 85017-77-2
Molecular Formula: Br2Co.xH2O
Formula Weight: 218.75(anhy)
Physical Form: Crystalline Granular
Density: 2.46
Sensitivity: Hygroscopic
Appearance: Red-purple
Odor: Odorless
Melting Point: 47-48° dec.
Merck Reference: 14,2435
MDL No.: MFCD00661152
Hygrometers, catalysts
Formula:
Br₂Co·H₂O Melting Pt: 47…48 °C (dec.) Density: 2.46 Storage Temperature: Ambient |
MDL Number:
MFCD00661152 CAS Number: 85017-77-2 EINECS: 232-166-7 Merck Index: 14,2435 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...