Copper(I) thiocyanate ≥96%

Supplier: Thermo Scientific Chemicals
Warning

40220-18 40220-30
CAAA40220-18EA 70.31 CAD
CAAA40220-18 CAAA40220-30
Copper(I) thiocyanate ≥96%
Copper (I) thiocyanate

MDL: MFCD00010980 Practically insoluble in water, alcohol. Soluble in NH4OH, ether


Purity: 96% min CAS No.: 1111-67-7 Molecular Formula: CuSCN Formula Weight: 121.63 Physical Form: Powder Density: 2.84 Melting Point: 1084° Solubility: Practically insoluble in water, alcohol. Soluble in NH4OH, ether Merck Reference: 14,2669 UN No.: 3077 MDL No.: MFCD00010980

Formula: CCuNS
Melting Pt: 1084 °C
Density: 2.84
Storage Temperature: Ambient
MDL Number: MFCD00010980
CAS Number: 1111-67-7
EINECS: 214-183-1
UN: 3077
ADR: 9,III
Merck Index: 14,2669

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR