Gallium nitride ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Gallium nitride, Gallium(III) nitride
40218-09
40218-18
CAAA40218-09EA
364.43
CAD
CAAA40218-09
CAAA40218-18
Gallium nitride ≥99.99% (metals basis)
Gallium nitride
MDL: MFCD00016108
Insoluble in water, dilute acids. Slightly soluble in hot conc. H2SO4, hot conc. NaOH
Purity: 99.99%
CAS No.: 25617-97-4
Molecular Formula: GaN
Formula Weight: 83.73
Physical Form: Powder
Density: 6.1
Appearance: Yellow
Melting Point: 800° subl.
Solubility: Insoluble in water, dilute acids. Slightly soluble in hot conc. H2SO4, hot conc. NaOH
Merck Reference: 14,4351
MDL No.: MFCD00016108
Formula:
GaN MW: 83.73 Melting Pt: 800 (subl.) Density: 6.1 Storage Temperature: Ambient |
MDL Number:
MFCD00016108 CAS Number: 25617-97-4 EINECS: 247-129-0 Merck Index: 14,4351 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...