Ammonium fluoride hydrate ≥99.9975% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

40216-14 40216-22
CAAA40216-14EA 268.6 CAD
CAAA40216-14 CAAA40216-22
Ammonium fluoride hydrate ≥99.9975% (metals basis), Puratronic®
Ammonium fluoride hydrate

MDL: MFCD00011423


Purity: 99.9975% CAS No.: 12125-01-8 Molecular Formula: H4FN.xH2O (x≈0.5) Formula Weight: 37.04(anhy) Physical Form: Crystalline Aggregates Appearance: White Odor: Odorless UN No.: 2505 MDL No.: MFCD00011423

Formula: NH₄F·H₂O
Melting Pt: 160 °C
Density: 1.0088…1.0096 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011423
CAS Number: 12125-01-8
EINECS: 235-185-9
UN: 2505
ADR: 6.1,III
Merck Index: 13,00524

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR