Chromium(III) telluride ≥99.5% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
40206-06
40206-14
CAAA40206-06EA
118.5
CAD
CAAA40206-06
CAAA40206-14
Chromium(III) telluride ≥99.5% (metals basis)
Chromium (III) telluride
MDL: MFCD00016040
Purity: 99.5%
CAS No.: 12053-39-3
Molecular Formula: Cr2Te3
Formula Weight: 486.79
Physical Form: -60+100 Mesh Powder
Density: 7.0
Appearance: Gray-black
Odor: Odorless
Melting Point: ca 1300°
MDL No.: MFCD00016040
Formula:
Cr₂Te₃ Melting Pt: ∼1300 °C Density: 7,0 Storage Temperature: Ambient |
MDL Number:
MFCD00016040 CAS Number: 12053-39-3 EINECS: 235-003-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...