Sodium iodide ≥99.5% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Hydriodic acid sodium salt, Sodium monoiodide, Ioduril
MDL: MFCD00003532
Soluble in water, alcohol, glycerol, acetone. Insoluble in ether
Fieser: 1,1087 4,456 6,543 7,338 10,365 12,449 13,282 14,289 21,404
Purity: 99.5% min
CAS No.: 7681-82-5
Molecular Formula: NaI
Formula Weight: 149.89
Physical Form: Crystalline Powder
Density: 3.67
Refractive Index: 1.7745
Sensitivity: Hygroscopic
Odor: Odorless
Melting Point: 651°
Boiling Point: 1300-1304°
Solubility: Soluble in water, alcohol, glycerol, acetone. Insoluble in ether
Merck Reference: 14,8631
MDL No.: MFCD00003532
Formula:
NaI Boiling Pt: 1300…1304 °C Melting Pt: 651 °C Density: 3.67 Storage Temperature: Ambient |
MDL Number:
MFCD00003532 CAS Number: 7681-82-5 EINECS: 231-679-3 UN: 3077 ADR: 9,III Merck Index: 14,8631 |
Specification Test Results
Insoluble matter | 0.01% |
Chloride and bromide (as Cl) | 0.01% |
IO3 | 2 ppm |
PO3 | 0.00001 |
SO3 | 0.005% |
Ba | 0.002% |
Ca | 0.001% |
Heavy metals (as Pb) | 5 ppm |
Fe | 5 ppm |
K | 0.01% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...