Ammonium cerium (IV) sulfate dihydrate ≥94% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
40194-30
40194-A1
40194-A7
CAAA40194-30EA
159.58
CAD
CAAA40194-30
CAAA40194-A1
CAAA40194-A7
Ammonium cerium (IV) sulfate dihydrate ≥94% ACS
Ammonium cerium (IV) sulphate dihydrate
MDL: MFCD00149417
Purity: 94% min
CAS No.: 10378-47-9
Molecular Formula: (NH4)4Ce(SO4)4.2H2O
Formula Weight: 632.56 (596.53anhy)
Physical Form: Crystalline
Appearance: Yellow to orange
Odor: Odorless
Melting Point: 130°
MDL No.: MFCD00149417
Formula:
(NH₄)₂Ce(SO₄)₄·2H₂O Melting Pt: 130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149417 CAS Number: 10378-47-9 EINECS: 231-567-4 |
Specification Test Results
Insoluble in dilute sulfuric acid | 0.05% |
PO3 | 0.003% |
Fe | 0.01% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...