Phosphorus(III) bromide ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Phosphorus tribromide

40191-14 40191-22
CAAA40191-14EA 84.53 CAD
CAAA40191-14 CAAA40191-22
Phosphorus(III) bromide ≥99% (metals basis)
Phosphorus (III) bromide

MDL: MFCD00011436 Beilstein Registry No.: 3903060 Soluble in acetone and carbon disulfide. Decomposes in water and alcohol


Purity: 99% CAS No.: 7789-60-8 Molecular Formula: PBr3 Formula Weight: 270.70 Physical Form: Liquid Density: 2.852 Refractive Index: 1.697 Sensitivity: Moisture Sensitive Odor: Pungent Melting Point: -40° Boiling Point: 172.9° Solubility: Soluble in acetone and carbon disulfide. Decomposes in water and alcohol Beilstein Reference: 3903060 Merck Reference: 14,7357 UN No.: 1808 MDL No.: MFCD00011436

Formula: PBr₃
MW: 270.69 g/mol
Boiling Pt: 172.9 °C
Melting Pt: –40 °C
Density: 2.852
Storage Temperature: Ambient
MDL Number: MFCD00011436
CAS Number: 7789-60-8
EINECS: 232-178-2
UN: 1808
ADR: 8,II
Merck Index: 14,7357

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR