Copper(I) oxide ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cuprous oxide, Dicopper oxide

40188-14 40188-22 40188-36
CAAA40188-14EA 48.66 CAD
CAAA40188-14 CAAA40188-22 CAAA40188-36
Copper(I) oxide ≥99% (metals basis)
Copper (I) oxide

MDL: MFCD00010974


Purity: 99% CAS No.: 1317-39-1 Molecular Formula: Cu2O Formula Weight: 143.08 Physical Form: -200 Mesh Powder Density: 6.0 Refractive Index: 2.705 Sensitivity: Air and Moisture Sensitive Appearance: Red to dark red Odor: Odorless Melting Point: 1235° Boiling Point: 1800° dec. Merck Reference: 14,2664 UN No.: 3077 MDL No.: MFCD00010974

Formula: Cu₂O
MW: 143.09 g/mol
Boiling Pt: 1800 (dec.)
Melting Pt: 1232
Density: 6.0
Storage Temperature: Ambient
MDL Number: MFCD00010974
CAS Number: 1317-39-1
EINECS: 215-270-7
UN: 3077
ADR: 9 III
Merck Index: 14,2664

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR