Silver(II) oxide ≥99.9% (metals basis) Ag ≥ 86.6%

Supplier: Thermo Scientific Chemicals
Danger

40187-06 40187-14 40187-22
CAAA40187-06EA 155.16 CAD
CAAA40187-06 CAAA40187-14 CAAA40187-22
Silver(II) oxide ≥99.9% (metals basis) Ag ≥ 86.6%
Silver (II) oxide

MDL: MFCD00044285


Purity: 99.9% CAS No.: 1301-96-8 Molecular Formula: AgO Formula Weight: 123.87 Physical Form: -100 Mesh Powder Density: 7.44 Appearance: Grey to black Odor: Odorless Melting Point: 100° dec. Merck Reference: 14,8522 UN No.: 3085 MDL No.: MFCD00044285

Formula: AgO
Melting Pt: >100 °C (dec.)
Density: 7.44
Storage Temperature: Ambient
MDL Number: MFCD00044285
CAS Number: 1301-96-8
EINECS: 215-098-2
UN: 3085
ADR: 5.1,II
Merck Index: 14,8522

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR