Indium(III) nitrate hydrate ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Indium trinitrate hydrate
40185-09
40185-18
CAAA40185-09EA
148.36
CAD
CAAA40185-09
CAAA40185-18
Indium(III) nitrate hydrate ≥99.99% (metals basis)
Indium (III) nitrate hydrate
MDL: MFCD00149742
Purity: 99.99%
CAS No.: 207398-97-8
Molecular Formula: InN3O9.xH2O
Formula Weight: 300.83(anhy)
Physical Form: Lump/Powder
Sensitivity: Hygroscopic
Appearance: Off-white
Melting Point: 100° -H2O
UN No.: 1477
MDL No.: MFCD00149742
Formula:
InN₃O₉·H₂O MW: 318.85 g/mol Melting Pt: 100 °C (-H₂O) Storage Temperature: Ambient |
MDL Number:
MFCD00149742 CAS Number: 207398-97-8 EINECS: 237-393-5 UN: 1477 ADR: 5.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...