Cobalt(II,III) oxide ≥99.7% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

40184-14 40184-22 40184-36 40184-A4
CAAA40184-14EA 70.15 CAD
CAAA40184-14 CAAA40184-22 CAAA40184-36 CAAA40184-A4
Cobalt(II,III) oxide ≥99.7% (metals basis)
Cobalt (II,III) oxide

Enamels, semiconductors, grinding wheels


Purity: 99.7% CAS No.: 1308-06-1 Molecular Formula: Co3O4 Formula Weight: 240.80 Physical Form: -400 Mesh Powder Density: 6.11 Appearance: Black Melting Point: 900° dec. Solubility: Soluble in acids and alkalies Merck Reference: 14,2429 UN No.: 3077 MDL No.: MFCD00010939


Enamels, semiconductors, grinding wheels

Formula: Co₃O₄
Melting Pt: 900 °C (dec.)
Density: 6.11
Storage Temperature: Ambient
MDL Number: MFCD00010939
CAS Number: 1308-06-1
EINECS: 215-157-2
UN: 3077
ADR: 9,III
Merck Index: 14,2429

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR