Cobalt ≥99.95% (metals basis), foil 0.025 mm (0.001 in), Thickness: 0.025 mm (0.001 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
40183-FF
40183-FI
CAAA40183-FFEA
322.32
CAD
CAAA40183-FF
CAAA40183-FI
Cobalt ≥99.95% (metals basis), foil 0.025 mm (0.001 in), Thickness: 0.025 mm (0.001 in)
Cobalt
MDL: MFCD00010935
Purity: 99.95%
CAS No.: 7440-48-4
Physical Form: ≈0.14g/25x25mm
Odor: Odorless
MDL No.: MFCD00010935
Formula:
Co MW: 58.93 g/mol Boiling Pt: 2870 Melting Pt: 1495 Density: 8.9 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010935 CAS Number: 7440-48-4 EINECS: 231-158-0 Merck Index: 12,02488 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...