N,O-Bis(trimethylsilyl)trifluoroacetamide with 10% trimethylchlorosilane
Supplier: Thermo Scientific Chemicals
|
Danger
|
40174-06
40174-14
CAAA40174-06EA
71.26
CAD
CAAA40174-06
CAAA40174-14
N,O-Bis(trimethylsilyl)trifluoroacetamide with 10% trimethylchlorosilane
N,O-Bis(trimethylsilyl)trifluoroacetamide with 10% trimethylchlorosilane
MDL: MFCD00008269
CAS No.: 25561-30-2
Molecular Formula: C8H18F3NOSi2
Formula Weight: 257.40
Storage Temperature: 2°C to 8°C
Flash Point: 23°(73°F)
Physical Form: Liquid
Sensitivity: Moisture Sensitive
Appearance: Colorless to pale yellow
Beilstein Reference: 3607620
UN No.: 2924
MDL No.: MFCD00008269
Formula:
C₈H₁₈F₃NOSi₂ Flash Pt: 23 °C (73 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008269 CAS Number: 25561-30-2 EINECS: 247-103-9 UN: 2924 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...