Tetrachlorosilane ≥99.998% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
40148-AC
40148-AE
CAAA40148-ACEA
89.74
CAD
CAAA40148-AC
CAAA40148-AE
Tetrachlorosilane ≥99.998% (metals basis)
Tetrachlorosilane
MDL: MFCD00011229
Purity: 99.998%
CAS No.: 10026-04-7
Molecular Formula: Cl4Si
Formula Weight: 169.90
Physical Form: Liquid
Density: 1.483
Appearance: Colorless
Odor: Acrid
Melting Point: -70°
Boiling Point: 57.6°
Merck Reference: 14,8500
UN No.: 1818
MDL No.: MFCD00011229
Formula:
Cl₄Si Boiling Pt: 57.6 °C Melting Pt: –70 °C Density: 1.483 Storage Temperature: Ambient |
MDL Number:
MFCD00011229 CAS Number: 10026-04-7 EINECS: 233-054-0 UN: 1818 ADR: 8,II Merck Index: 14,8500 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...