Sodium dihydrogen phosphate ≥96%, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

40145-30 40145-A1
CAAA40145-30EA 80.9 CAD
CAAA40145-30 CAAA40145-A1
Sodium dihydrogen phosphate ≥96%, Technical Grade
Sodium dihydrogen phosphate

MDL: MFCD00003527 Freely soluble in water. Insoluble in alcohol


Purity: 96% CAS No.: 7558-80-7 Molecular Formula: H2NaO4P Formula Weight: 119.98 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Odor: Odorless Solubility: Freely soluble in water. Insoluble in alcohol Merck Reference: 14,8660 MDL No.: MFCD00003527

Formula: NaH₂PO₄
Melting Pt: ∼650 °C
Density: 0.9996 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003527
CAS Number: 7558-80-7
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 14,8660

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR