Calcium sulfate, anhydrous 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Calcium sulphate (anhydrous) anhydrite, Gypsum, Calcium sulfate (anhydrous) anhydrite
40144-30
40144-A1
40144-A7
CAAA40144-30EA
60.2
CAD
CAAA40144-30
CAAA40144-A1
CAAA40144-A7
Calcium sulfate, anhydrous 99%
Calcium sulphate
Drying agent, laboratory desiccant, paper filler
Purity: 99%
CAS No.: 7778-18-9
Molecular Formula: CaO4S
Formula Weight: 136.14
Physical Form: Powder
Density: 2.960
Appearance: White
Odor: Odorless
Melting Point: 1450°
Merck Reference: 14,1706
MDL No.: MFCD00010912
Drying agent, laboratory desiccant, paper filler
Formula:
CaO₄S MW: 136.14 g/mol Melting Pt: 1350 Density: 2.960 Storage Temperature: Ambient |
MDL Number:
MFCD00010912 CAS Number: 7778-18-9 EINECS: 231-900-3 Merck Index: 14,1706 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...