Calcium iodide hydrate ≥98%

Supplier: Thermo Scientific Chemicals
Warning

40136-22 40136-36
CAAA40136-22EA 152.31 CAD
CAAA40136-22 CAAA40136-36
Calcium iodide hydrate ≥98%
Calcium iodide hydrate

MDL: MFCD00149622 Freely soluble in water and alcohol


CAS No.: 71626-98-7 Molecular Formula: CaI2.xH2O Formula Weight: 293.89(anhy) Physical Form: Lump Density: 2.55 Sensitivity: Hygroscopic Appearance: White to grey Melting Point: 42° dec. Solubility: Freely soluble in water and alcohol Merck Reference: 14,1677 MDL No.: MFCD00149622

Formula: CaI₂·H₂O
MW: 311.9 g/mol
Melting Pt: 42 °C (dec.)
Density: 2.55
Storage Temperature: Ambient
MDL Number: MFCD00149622
CAS Number: 71626-98-7
EINECS: 233-276-8
Merck Index: 14,1677

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR