Indium(III) bromide, anhydrous ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

40126-09 40126-18
CAAA40126-09EA 252.8 CAD
CAAA40126-09 CAAA40126-18
Indium(III) bromide, anhydrous ≥99.99% (metals basis)
Indium (III) bromide

MDL: MFCD00011057


Purity: 99.99% CAS No.: 13465-09-3 Molecular Formula: Br3In Formula Weight: 354.55 Physical Form: Crystalline powder Density: 4.74 Appearance: White to yellow Odor: Odorless Melting Point: ca 436° MDL No.: MFCD00011057

Formula: Br₃In
Boiling Pt: 656 °C (1013 hPa)
Melting Pt: ∼ 436 °C
Density: 4.74
Storage Temperature: Ambient
MDL Number: MFCD00011057
CAS Number: 13465-09-3
EINECS: 236-692-8
UN: 3260
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR