Cerium(IV) oxide 20% in water, colloidal dispersion

Supplier: Thermo Scientific Chemicals

40125-22 40125-36 40125-A3
CAAA40125-22EA 72.21 CAD
CAAA40125-22 CAAA40125-36 CAAA40125-A3
Cerium(IV) oxide 20% in water, colloidal dispersion
Cerium (IV) oxide

MDL: MFCD00010933


CAS No.: 1306-38-3 Molecular Formula: CeO2 mol/mol 0.4 acetate, pH 3.0 Formula Weight: 172.12 Physical Form: 0.01-0.02 Micron Particles, in Liquid Density: 1.22 Odor: Odorless MDL No.: MFCD00010933

Formula: CeO₂
Density: 1.22
Storage Temperature: Ambient
MDL Number: MFCD00010933
CAS Number: 1306-38-3
EINECS: 215-150-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR