Indium(III) fluoride, anhydrous ≥96%

Supplier: Thermo Scientific Chemicals
Warning

40118-04 40118-09
CAAA40118-04EA 73.31 CAD
CAAA40118-04 CAAA40118-09
Indium(III) fluoride, anhydrous ≥96%
Indium (III) fluoride

MDL: MFCD00016149 Slightly soluble in water (0.04g/100ml). Freely soluble in dilute acids


Purity: 96% CAS No.: 7783-52-0 Molecular Formula: InF3 Formula Weight: 171.82 Physical Form: Powder Density: 4.39 Appearance: White Odor: Odorless Melting Point: ca 1170° Boiling Point: 1200° Solubility: Slightly soluble in water (0.04g/100ml). Freely soluble in dilute acids Merck Reference: 14,4959 MDL No.: MFCD00016149

Formula: F₃In
Boiling Pt: >1200 °C
Melting Pt: ∼ 1170 °C
Density: 4.39
Storage Temperature: Ambient
MDL Number: MFCD00016149
CAS Number: 7783-52-0
EINECS: 232-005-0
UN: 2923
ADR: 8,II
Merck Index: 14,4959

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR