Silver chromate ≥99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
40107-14
40107-22
CAAA40107-14EA
369.72
CAD
CAAA40107-14
CAAA40107-22
Silver chromate ≥99%
Silver chromate
MDL: MFCD00003402
Slightly soluble in water. Soluble in HNO3 and ammonia
Purity: 99% min
CAS No.: 7784-01-2
Molecular Formula: Ag2CrO4
Formula Weight: 331.73
Physical Form: Powder
Density: 5.63
Solubility: Slightly soluble in water. Soluble in HNO3 and ammonia
Merck Reference: 14,8510
UN No.: 1479
MDL No.: MFCD00003402
Formula:
Ag₂CrO₄ Boiling Pt: 1550 °C (1013 hPa) Density: 5.63 Storage Temperature: Ambient |
MDL Number:
MFCD00003402 CAS Number: 7784-01-2 EINECS: 232-043-8 UN: 1479 ADR: 5.1,III Merck Index: 14,8510 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...