Zinc sulfide ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Zinc (II) sulphide, Zinc (II) sulfide

40091-09 40091-18 40091-30
CAAA40091-09EA 45.82 CAD
CAAA40091-09 CAAA40091-18 CAAA40091-30
Zinc sulfide ≥99.99% (metals basis)
Zinc sulphide

MDL: MFCD00011301


Purity: 99.99% CAS No.: 1314-98-3 Molecular Formula: SZn Formula Weight: 97.43 Physical Form: -325 Mesh Powder Density: 4.10 Appearance: White Odor: Odorless Melting Point: 1700° Merck Reference: 14,10160 MDL No.: MFCD00011301

Formula: SZn
MW: 97.46 g/mol
Melting Pt: 1064
Density: 4.10
Storage Temperature: Ambient
MDL Number: MFCD00011301
CAS Number: 1314-98-3
EINECS: 215-251-3
Merck Index: 14,10160

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR