Indium(I) chloride, anhydrous ≥99.995% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
40033-02
40033-06
40033-14
CAAA40033-02EA
53.88
CAD
CAAA40033-02
CAAA40033-06
CAAA40033-14
Indium(I) chloride, anhydrous ≥99.995% (metals basis)
Indium (I) chloride
MDL: MFCD00148875
Purity: 99.995%
CAS No.: 13465-10-6
Molecular Formula: InCl
Formula Weight: 150.27
Physical Form: Crystalline, Packaged under argon
Density: 4.19
Melting Point: ca 225°
Boiling Point: 608°
UN No.: 3260
MDL No.: MFCD00148875
Formula:
InCl Boiling Pt: 608 °C Melting Pt: ∼225 °C Density: 4.19 Storage Temperature: Ambient |
MDL Number:
MFCD00148875 CAS Number: 13465-10-6 EINECS: 236-693-3 UN: 3260 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...