Potassium hexafluorophosphate ≥99%

Supplier: Thermo Scientific Chemicals
Danger

40030-22 40030-30 40030-36
CAAA40030-22EA 123.24 CAD
CAAA40030-22 CAAA40030-30 CAAA40030-36
Potassium hexafluorophosphate ≥99%
Potassium hexafluorophosphate

MDL: MFCD00011412


Purity: 99% min CAS No.: 17084-13-8 Molecular Formula: KPF6 Formula Weight: 184.07 Physical Form: Crystalline Density: 2.55 Appearance: White Odor: Odorless pH: 3.0-7.0 Melting Point: 575° UN No.: 3260 MDL No.: MFCD00011412

Formula: F₆KP
Melting Pt: 575 °C
Density: 2,55
Storage Temperature: Ambient
MDL Number: MFCD00011412
CAS Number: 17084-13-8
EINECS: 241-143-0
UN: 3260
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR