Triethyl phosphate 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
40001-14
40001-22
40001-36
CAAA40001-14EA
30.34
CAD
CAAA40001-14
CAAA40001-22
CAAA40001-36
Triethyl phosphate 99+%
Triethyl phosphate
MDL: MFCD00009077
Purity: 99+%
CAS No.: 78-40-0
Molecular Formula: (C2H5O)3P(O)
Formula Weight: 182.16
Flash Point: 115°(240°F)
Physical Form: Liquid
Density: 1.072
Refractive Index: 1.4050
Odor: Mild
Boiling Point: 215°
Merck Reference: 14,9674
MDL No.: MFCD00009077
Formula:
C₆H₁₅O₄P Boiling Pt: 215 °C Melting Pt: –57 °C Density: 1.072 Flash Pt: 115 °C (240 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009077 CAS Number: 78-40-0 EINECS: 201-114-5 Merck Index: 14,9674 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...