Osmium ≥99.95% (metals basis), powder -20 mesh

Supplier: Thermo Scientific Chemicals
Danger

39998-03 39998-06
CAAA39998-03EA 306.52 CAD
CAAA39998-03 CAAA39998-06
Osmium ≥99.95% (metals basis), powder -20 mesh
Osmium

MDL: MFCD00011147


Purity: 99.95% CAS No.: 7440-04-2 Odor: Strong UN No.: 3089 MDL No.: MFCD00011147

Formula: Os
MW: 190.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011147
CAS Number: 7440-04-2
EINECS: 231-114-0
UN: 3089
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR