Gold ≥99.96% (metals basis), spherical powder APS 1,5-3,0 µm
Supplier: Thermo Scientific Chemicals
39818-03
39818-06
39818-03
39818-MD
39818-MD
39818-06
CAAA39818-03EA
453.46
CAD
CAAA39818-03
AA39818-03
CAAA39818-06
CAAA39818-MD
AA39818-MD
AA39818-06
Gold ≥99.96% (metals basis), spherical powder APS 1,5-3,0 µm
Gold
Formula:
Au MW: 196.97 g/mol Boiling Pt: 2800 °C (1013 hPa) Melting Pt: 1063 °C Density: 19.3 g/cm³ (20 °C) |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 Merck Index: 12,04526 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...