Aluminium oxide, γ-phase ≥99.97% (metals basis)

Supplier: Thermo Scientific Chemicals

39812-14 39812-22 39812-36
CAAA39812-14EA 80.26 CAD
CAAA39812-14 CAAA39812-22 CAAA39812-36
Aluminium oxide, γ-phase ≥99.97% (metals basis)
Aluminium oxide, γ-phase

MDL: MFCD00003424


Purity: 99.97% CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 3 Micron APS Powder, S.A. 80-120m2/g Odor: Odorless Merck Reference: 14,356 MDL No.: MFCD00003424

Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980 °C (1013 hPa)
Melting Pt: 2045 °C
Density: 3.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
Merck Index: 14,356

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR