EDTA disodium salt standardised solution 0.050 M (0.100 N) in aqueous solution

Supplier: Thermo Scientific Chemicals

Synonyms: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt

39781-K2 39781-K7
CAAA39781-K2EA 82.95 CAD
CAAA39781-K2 CAAA39781-K7
EDTA disodium salt standardised solution 0.050 M (0.100 N) in aqueous solution
EDTA disodium salt

MDL: MFCD00070672


CAS No.: 139-33-3 Molecular Formula: (NaO2CCH2)2NCH2CH2N(CH2CO2H)2 Formula Weight: 336.22 Physical Form: Liquid Odor: Odorless MDL No.: MFCD00070672

Formula: C₁₀H₁₄N₂Na₂O₈
Storage Temperature: Ambient
MDL Number: MFCD00070672
CAS Number: 139-33-3
EINECS: 205-358-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR