Tungsten ≥99.9% (metals basis), powder -325 mesh

Supplier: Thermo Scientific Chemicals
Warning

39749-22 39749-36 39749-A3
CAAA39749-22EA 99.38 CAD
CAAA39749-22 CAAA39749-36 CAAA39749-A3
Tungsten ≥99.9% (metals basis), powder -325 mesh
Tungsten

MDL: MFCD00011461


Purity: 99.9% CAS No.: 7440-33-7 Appearance: Grey UN No.: 3089 MDL No.: MFCD00011461

Formula: W
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
UN: 3089
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR