Fullerene C60 ≥99.5%, powder
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Fullerene - C60, Buckminster-Fullerene C60, C60 Fullerene, Fullerene-C60, Bucky Ball, Footballen
39722-03
39722-MC
CAAA39722-03EA
579.86
CAD
CAAA39722-03
CAAA39722-MC
Fullerene C60 ≥99.5%, powder
Fullerene C60
MDL: MFCD00151408
Purity: 99.5%
CAS No.: 99685-96-8
Density: 1.7
Appearance: Black/brown
Boiling Point: 500-600° subl.
UN No.: 1325
MDL No.: MFCD00151408
Formula:
C₆₀ Boiling Pt: 500…600 °C (subl.) Melting Pt: > 280 °C Density: 1.7 Storage Temperature: Ambient |
MDL Number:
MFCD00151408 CAS Number: 99685-96-8 UN: 1325 ADR: 4.1,III Merck Index: 13,01447 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...