Carbon ≥99.96%, yarn, woven from 0.076 mm (0.003 in) dia fibers

Supplier: Thermo Scientific Chemicals

Synonyms: Charcoal

39721-G5 39721-G9
CAAA39721-G5EA 56.25 CAD
CAAA39721-G5 CAAA39721-G9
Carbon ≥99.96%, yarn, woven from 0.076 mm (0.003 in) dia fibers
Carbon

MDL: MFCD00133992


MDL No.: MFCD00133992

Formula: C
Melting Pt: 3727 °C
Density: 3.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR