Copper, gauze woven from wire 50 mesh, Ø 0.23 mm (0.009 in)

Supplier: Thermo Scientific Chemicals

39713-WP 39713-WQ
CAAA39713-WPEA 106.33 CAD
CAAA39713-WP CAAA39713-WQ
Copper, gauze woven from wire 50 mesh, Ø 0.23 mm (0.009 in)
Copper

MDL: MFCD00010965 Notes: Open area: 30.3%; Width of opening: 0.011in


CAS No.: 7440-50-8 Physical Form: Wire Cloth Odor: Odorless MDL No.: MFCD00010965

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR