Nickel, gauze woven from wire 20 mesh, Ø 0.18 mm (0.007 in)

Supplier: Thermo Scientific Chemicals
Danger

39704-0S 39704-WO
CAAA39704-0SEA 393.42 CAD
CAAA39704-0S CAAA39704-WO
Nickel, gauze woven from wire 20 mesh, Ø 0.18 mm (0.007 in)
Nickel

MDL: MFCD00011137 Notes: Open area: 74%; Width of opening: 0.043in


CAS No.: 7440-02-0 Physical Form: Wire Cloth Odor: Odorless MDL No.: MFCD00011137

Formula: Ni
Boiling Pt: 3007 °C (1013 hPa)
Melting Pt: 1453 °C
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,III
Merck Index: 12,06582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR