Nickel, gauze woven from wire 40 mesh, Ø 0.13 mm (0.005 in)

Supplier: Thermo Scientific Chemicals
Danger

39703-HX 39703-WM
CAAA39703-HXEA 349.18 CAD
CAAA39703-HX CAAA39703-WM
Nickel, gauze woven from wire 40 mesh, Ø 0.13 mm (0.005 in)
Nickel

MDL: MFCD00011137 Notes: Open area: 64%; Width of opening: 0.02in


CAS No.: 7440-02-0 Physical Form: Wire Cloth Odor: Odorless MDL No.: MFCD00011137

Formula: Ni
Boiling Pt: 3007 °C (1013 hPa)
Melting Pt: 1453 °C
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,III
Merck Index: 12,06582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR