Silver 99,999 Metals basis, needles

Supplier: Thermo Scientific Chemicals

39697-09 39697-18
CAAA39697-09EA 241.74 CAD
CAAA39697-09 CAAA39697-18
Silver 99,999 Metals basis, needles
Silver

MDL: MFCD00003397


Purity: 99.999% CAS No.: 7440-22-4 Odor: Characteristic UN No.: 3077 MDL No.: MFCD00003397

Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
UN: 3077
ADR: 9 III
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR