Vanadium ≥99.7% (metals basis), granules 1-3 mm (0.04-0.12 in) for vacuum deposition
Supplier: Thermo Scientific Chemicals
39693-09
39693-18
39693-30
CAAA39693-09EA
135.56
CAD
CAAA39693-09
CAAA39693-18
CAAA39693-30
Vanadium ≥99.7% (metals basis), granules 1-3 mm (0.04-0.12 in) for vacuum deposition
Vanadium
MDL: MFCD00011453
Purity: 99.7%
CAS No.: 7440-62-2
Physical Form: For vacuum deposition
Odor: Odorless
MDL No.: MFCD00011453
Formula:
V MW: 50.94 g/mol Boiling Pt: 3477 Melting Pt: 1896 Density: 6.015 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011453 CAS Number: 7440-62-2 EINECS: 231-171-1 Merck Index: 12,10053 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...