Tungsten ≥99.999% (metals basis), powder, Puratronic® APS 4-6 µm (max. 1ppm Mo)

Supplier: Thermo Scientific Chemicals
Warning

39691-09 39691-18 39691-04
CAAA39691-09EA 322.32 CAD
CAAA39691-09 CAAA39691-18 CAAA39691-04
Tungsten ≥99.999% (metals basis), powder, Puratronic® APS 4-6 µm (max. 1ppm Mo)
Tungsten

MDL: MFCD00011461


Purity: 99.999% CAS No.: 7440-33-7 Appearance: Grey UN No.: 3089 MDL No.: MFCD00011461

Formula: W
MW: 183.84 g/mol
Boiling Pt: 5660
Melting Pt: 3410
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
UN: 3089
ADR: 4.1 III
Merck Index: 12,09945

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR