Molybdenum ≥99.9% (metals basis), powder APS ∼2-4 µm

Supplier: Thermo Scientific Chemicals
Warning

39687-14 39687-22
CAAA39687-14EA 86.74 CAD
CAAA39687-14 CAAA39687-22
Molybdenum ≥99.9% (metals basis), powder APS ∼2-4 µm
Molybdenum

MDL: MFCD00003465 Notes: From Hydrogen reduced oxide


Purity: 99.9% CAS No.: 7439-98-7 Appearance: Grey Odor: Odorless UN No.: 3089 MDL No.: MFCD00003465

Formula: Mo
Boiling Pt: 5560 °C (1013 hPa)
Melting Pt: 2622 °C
Density: 10,2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
UN: 3089
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR