Molybdenum ≥99.998% (metals basis excluding W), powder, Puratronic® -325 mesh (W 130 ppm)

Supplier: Thermo Scientific Chemicals
Warning

39686-14 39686-22
CAAA39686-14EA 385.52 CAD
CAAA39686-14 CAAA39686-22
Molybdenum ≥99.998% (metals basis excluding W), powder, Puratronic® -325 mesh (W 130 ppm)
Molybdenum

MDL: MFCD00003465


Purity: 99.999% CAS No.: 7439-98-7 Appearance: Grey Odor: Odorless UN No.: 3089 MDL No.: MFCD00003465

Formula: Mo
Boiling Pt: 5560 °C (1013 hPa)
Melting Pt: 2622 °C
Density: 10.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
UN: 3089
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR