Hafnium ≥99.9% (metals basis excluding Zr), pieces 3-12 mm (0.12-0.47 in) (Zr ∼ 3%)

Supplier: Thermo Scientific Chemicals

39683-09 39683-18
CAAA39683-09EA 221.2 CAD
CAAA39683-09 CAAA39683-18
Hafnium ≥99.9% (metals basis excluding Zr), pieces 3-12 mm (0.12-0.47 in) (Zr ∼ 3%)
Hafnium

MDL: MFCD00011032


Purity: 99.9% CAS No.: 7440-58-6 Odor: Odorless MDL No.: MFCD00011032

Formula: Hf
Storage Temperature: Ambient
MDL Number: MFCD00011032
CAS Number: 7440-58-6
EINECS: 231-166-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR