Copper ≥99.999% (metals basis), powder -100 mesh

Supplier: Thermo Scientific Chemicals
Warning

39682-06 39682-14 39682-22
CAAA39682-06EA 148.36 CAD
CAAA39682-06 CAAA39682-14 CAAA39682-22
Copper ≥99.999% (metals basis), powder -100 mesh
Copper

MDL: MFCD00010965


Purity: 99.999% CAS No.: 7440-50-8 Appearance: Red brown Odor: Odorless UN No.: 3089 MDL No.: MFCD00010965

Formula: Cu
Boiling Pt: 2595 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
UN: 3089
ADR: 4.1,III
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR