Titanium ≥99.99% (metals basis excluding Na, K), granules

Supplier: Thermo Scientific Chemicals
Warning

39677-18 39677-30
CAAA39677-18EA 188.02 CAD
CAAA39677-18 CAAA39677-30
Titanium ≥99.99% (metals basis excluding Na, K), granules
Titanium

MDL: MFCD00011264


Purity: 99.99% CAS No.: 7440-32-6 Odor: Odorless MDL No.: MFCD00011264

Formula: Ti
Boiling Pt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR