Strontium aluminate ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals

39647-14 39647-22
CAAA39647-14EA 140.94 CAD
CAAA39647-14 CAAA39647-22
Strontium aluminate ≥99.5% (metals basis)
Strontium aluminate

MDL: MFCD00054054


Purity: 99.5% CAS No.: 12004-37-4 Molecular Formula: SrAl2O4 Formula Weight: 205.58 Physical Form: -100 Mesh Powder Density: 3.559 Appearance: Silver Odor: Odorless MDL No.: MFCD00054054

Formula: Al₂O₄Sr
Density: 3,559
Storage Temperature: Ambient
MDL Number: MFCD00054054
CAS Number: 12004-37-4
EINECS: 234-455-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR