Dimethyldiphenyltin
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Dimethyldiphenylstannane
39617-03
39617-06
39617-14
CAAA39617-03EA
78.53
CAD
CAAA39617-03
CAAA39617-06
CAAA39617-14
Dimethyldiphenyltin
Dimethyldiphenyltin
MDL: MFCD00058812
CAS No.: 1080-43-9
Molecular Formula: (CH3)2Sn(C6H5)2
Formula Weight: 302.97
Physical Form: Liquid
UN No.: 2788
MDL No.: MFCD00058812
Formula:
C₁₄H₁₆Sn Storage Temperature: Ambient |
MDL Number:
MFCD00058812 CAS Number: 1080-43-9 UN: 2788 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...