Tributyltin fluoride

Supplier: Thermo Scientific Chemicals
Danger

39602-06 39602-14
CAAAA39602-06EA 79 CAD
CAAAA39602-06 CAAA39602-14
Tributyltin fluoride
Tributyltin fluoride

MDL: MFCD00013547


CAS No.: 1983-10-4 Molecular Formula: [CH3(CH2)3]3SnF Formula Weight: 309.04 Physical Form: Powder Sensitivity: Moisture Sensitive Odor: Pungent UN No.: 3146 MDL No.: MFCD00013547

Formula: C₁₂H₂₇FSn
Melting Pt: 248…252 °C
Storage Temperature: Ambient
MDL Number: MFCD00013547
CAS Number: 1983-10-4
EINECS: 217-847-9
UN: 3146
ADR: 6.1,III

Order Now

Specification Test Results

Form Powder

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR